Learn More
STEMCELL Technologies Fasudil (Dihydrochloride), Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 73662
Encompass
Inhibit Rho-associated coiled-coil containing protein kinase 2 (ROCK2) with Fasudil. Fasudil is used to improve adipocyte differentiation, preventing development of diabetes and nephropathy in insulin-resistant diabetic rats (Kikuchi et al. J Endocrinol, 2007). This ROCK pathway inhibitor reduces pulmonary arterial hypertension in rats (Oka et al. Circ Res, 2007); enhances neurological recovery after traumatic spinal cord injury (Hara et al. J Neurosurg, 2000); and inhibits corneal neovascularization after alkali burns and promotes the healing of corneal epithelial defects in mice (Zeng et al. Mol Vis, 2015). Fasudil is supplied as the dihydrochloride salt of the molecule with ≥98% purity; Molecular weight 581.5 g/mol; CAS Number 548472-68-0. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-6110