Learn More
Apexbio Technology LLC Fasudil (HA-1077) HCl 105628-07-7 200mg
SDP
Catalog No. 5000093225
Encompass_Preferred
Fasudil HA-1077 HCl (CAS 105628-07-7) is a small-molecule inhibitor targeting Rho-associated protein kinase (ROCK) a serine/threonine kinase It is designed to inhibit ROCK thereby regulating cellular processes such as proliferation migration and apoptosis Fasudil HA-1077 HCl exerts its biological activity primarily through ROCK inhibition In cell-based studies Fasudil inhibits proliferation and migration and promotes apoptosis in cancer cell models including bladder carcinoma (5637 UM-UC-3) and oral squamous cell carcinoma (SCC-4) its ROCK inhibitory activity shows an IC50 value of approximately 0 74 M Based on these pharmacological properties Fasudil HA-1077 HCl holds research potential in oncology and disease models involving ROCK-mediated signaling
Catalog No. 50-000-93225 Supplier Apexbio Technology LLC Supplier No. A5734200
Fasudil HA-1077 HCl (CAS 105628-07-7) is a small-molecule inhibitor targeting Rho-associated protein kinase (ROCK) a serine/threonine kinase It is designed to inhibit ROCK thereby regulating cellular processes such as proliferation migration and apoptosis Fasudil HA-1077 HCl exerts its biological activity primarily through ROCK inhibition In cell-based studies Fasudil inhibits proliferation and migration and promotes apoptosis in cancer cell models including bladder carcinoma (5637 UM-UC-3) and oral squamous cell carcinoma (SCC-4) its ROCK inhibitory activity shows an IC50 value of approximately 0 74 M Based on these pharmacological properties Fasudil HA-1077 HCl holds research potential in oncology and disease models involving ROCK-mediated signaling