Learn More
Selleck Chemical LLC Fasudil HCl S1573-1g
SDP
Catalog No. 503111299
Encompass_Preferred
Fasudil HCl a potent and selective inhibitor of Rho kinase displays less potent inhibiton over PKA PKG PKC and MLCK with Ki of 1 6 1 6 3 3 and 36 M in cell-free assays respectively Fasudil is also a calcium channel blocker
Catalog No. 50-311-1299 Supplier Selleck Chemical LLC Supplier No. S15731G
Add to Cart
Add to Cart
Fasudil HCl a potent and selective inhibitor of Rho kinase displays less potent inhibiton over PKA PKG PKC and MLCK with Ki of 1 6 1 6 3 3 and 36 M in cell-free assays respectively Fasudil is also a calcium channel blocker