Learn More
Medchemexpress LLC HY-10341 500mg , Fasudil (Hydrochloride) HA-1077;AT-877;Fasudil HCl CAS:105628-07-7 Purity:98%
SDP
Catalog No. 501492399
Encompass_Preferred
Medchemexpress, HY-10341 500mg Fasudil (Hydrochloride) HA-1077;AT-877;Fasudil HCl CAS:105628-07-7 Formula:C14H18ClN3O2S Ki: 0.33 μM (ROCK1), 1.0 μM (PKA), 9.3 μM (PKC), 55 μM (MLCK) [8] Purity:98% Fasudil Hydrochloride is a potent inhibitor of ROCK1 , PKA , PKC , and MLCK with K i of 0.33 μM, 1.0 μM, 9.3 μM and 55 μM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2399 Supplier Medchemexpress LLC Supplier No. HY10341500MG
Medchemexpress, HY-10341 500mg Fasudil (Hydrochloride) HA-1077;AT-877;Fasudil HCl CAS:105628-07-7 Formula:C14H18ClN3O2S Ki: 0.33 μM (ROCK1), 1.0 μM (PKA), 9.3 μM (PKC), 55 μM (MLCK) [8] Purity:98% Fasudil Hydrochloride is a potent inhibitor of ROCK1 , PKA , PKC , and MLCK with K i of 0.33 μM, 1.0 μM, 9.3 μM and 55 μM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.