Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Fasudil hydrochloride | 105628-07-7 | MFCD00943198 | 100.0% | 327.83 g/mol | C14H18ClN3O2S | 1 G
SDP

Catalog No. 5000246667
Encompass_Preferred

Fasudil hydrochloride is the hydrochloride salt of fasudil, a small-molecule Rho kinase (ROCK) inhibitor used in biochemical and cellular research to probe cytoskeletal dynamics, vascular tone, and related signaling pathways. It is provided as a high-purity solid suitable for laboratory research.

  • RhoA/ROCK inhibitor for biochemical and cellular studies.
  • CAS number 105628-07-7 and molecular weight 327.83 g/mol.
  • Molecular formula C14H18ClN3O2S.
  • High purity (~99.97%) suitable for research use.
  • Solid, white to off-white appearance; soluble in DMSO and water.
  • Recommended storage: 4°C sealed; in solvent: -80°C (1 year) or -20°C (6 months).
  • Offered in laboratory pack sizes including 1 g.

Catalog No. 50-002-46667 Supplier Medchemexpress LLC Supplier No. HY103411G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fasudil hydrochloride is the hydrochloride salt of fasudil, a small-molecule Rho kinase (ROCK) inhibitor used in biochemical and cellular research to probe cytoskeletal dynamics, vascular tone, and related signaling pathways. It is provided as a high-purity solid suitable for laboratory research.

  • RhoA/ROCK inhibitor for biochemical and cellular studies.
  • CAS number 105628-07-7 and molecular weight 327.83 g/mol.
  • Molecular formula C14H18ClN3O2S.
  • High purity (~99.97%) suitable for research use.
  • Solid, white to off-white appearance; soluble in DMSO and water.
  • Recommended storage: 4°C sealed; in solvent: -80°C (1 year) or -20°C (6 months).
  • Offered in laboratory pack sizes including 1 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.