Learn More
Medchemexpress LLC Fasudil hydrochloride | 105628-07-7 | MFCD00943198 | 100.0% | 327.83 g/mol | C14H18ClN3O2S | 10 G
SDP
Catalog No. 5000244654
Encompass_Preferred
Fasudil hydrochloride is the hydrochloride salt of fasudil, a small-molecule inhibitor of RhoA/ROCK kinases used in biochemical and cell-based research. It is supplied for analytical and research applications and is typically handled as a solid powder with known solubility in DMSO and water.
Catalog No. 50-002-44654 Supplier Medchemexpress LLC Supplier No. HY1034110G
Fasudil hydrochloride is the hydrochloride salt of fasudil, a small-molecule inhibitor of RhoA/ROCK kinases used in biochemical and cell-based research. It is supplied for analytical and research applications and is typically handled as a solid powder with known solubility in DMSO and water.