Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Fat Diet, High, Modified

Catalog No. 0296019250 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Packaging Quantity:
20 kg
50 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
02-960-19250 50 kg
02-960-19220 20 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-960-19250 Supplier MP Biomedicals™ Supplier No. 0296019250
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Fat Diet, High, Modified is a diet for studying obesity in animal models.

  • Normal High Fat Diet modified to contain essential trace minerals, DL-Methionine and Fiber.
  • Fat content is 45% fat (product must be stored before use).
  • Typically provides 5.81 kcal/gm diet.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Fat Diet
Format Pellets
Packaging Quantity 50 kg

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.