Learn More
New England Biolabs, Inc. FatI – 250 units
SDP
Catalog No. 50995273
Encompass
A restriction endonuclease that recognizes the sequence ^CATG_.
Catalog No. 50-995-273 Supplier New England Biolabs, Inc. Supplier No. R0650L
A restriction endonuclease that recognizes the sequence ^CATG_.