Learn More
New England Biolabs, Inc. FatI – 50 units
SDP
Catalog No. 50995274
Encompass
A restriction endonuclease that recognizes the sequence ^CATG_.
Catalog No. 50-995-274 Supplier New England Biolabs, Inc. Supplier No. R0650S
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence ^CATG_.