Learn More
Medchemexpress LLC Fatostatin (125B11) | 125256-00-0 | 98.2% | C18H18N2S | 200 MG
SDP
Catalog No. 5000355328
Encompass_Preferred
Fatostatin (125B11) is a specific inhibitor of SREBP activation that impairs the activation of SREBP-1 and SREBP-2. It binds to SCAP (SREBP cleavage-activating protein) and inhibits the ER-Golgi translocation of SREBPs. Fatostatin decreases the transcription of lipogenic genes in cells. It also possesses antitumor properties and lowers hyperglycemia in ob/ob mice.
  • Specific inhibitor of SREBP activation.
  • Impairs activation of SREBP-1 and SREBP-2.
  • Binds to SCAP and inhibits ER-Golgi translocation of SREBPs.
  • Decreases transcription of lipogenic genes in cells.
  • Possesses antitumor properties.
  • Lowers hyperglycemia in ob/ob mice.
  • Inhibits androgen-independent prostate cancer cell proliferation.
  • Inhibits insulin-induced adipogenesis of 3T3-L1 cells.
  • Reduced adiposity and ameliorated fatty liver in ob/ob mice.
Catalog No. 50-003-55328 Supplier Medchemexpress LLC Supplier No. HY14452200MG
Fatostatin (125B11) is a specific inhibitor of SREBP activation that impairs the activation of SREBP-1 and SREBP-2. It binds to SCAP (SREBP cleavage-activating protein) and inhibits the ER-Golgi translocation of SREBPs. Fatostatin decreases the transcription of lipogenic genes in cells. It also possesses antitumor properties and lowers hyperglycemia in ob/ob mice.
  • Specific inhibitor of SREBP activation.
  • Impairs activation of SREBP-1 and SREBP-2.
  • Binds to SCAP and inhibits ER-Golgi translocation of SREBPs.
  • Decreases transcription of lipogenic genes in cells.
  • Possesses antitumor properties.
  • Lowers hyperglycemia in ob/ob mice.
  • Inhibits androgen-independent prostate cancer cell proliferation.
  • Inhibits insulin-induced adipogenesis of 3T3-L1 cells.
  • Reduced adiposity and ameliorated fatty liver in ob/ob mice.