Learn More
Selleck Chemical LLC Fatostatin-5mg
SDP
Supplier:  Selleck Chemical LLC S97855MG
Encompass_Preferred
Fatostatin (125B11), a diarylthiazole derivative, is a specific inhibitor of Sterol regulatory element binding proteins (SREBPs) activation. Fatostatin binds to SCAP (SREBP cleavage-activating protein), and inhibits the ER-Golgi translocation of SREBPs. Fatostatin suppresses growth and enhances apoptosis in cancer cells.
Catalog No. 50-238-9610