Learn More
Medchemexpress LLC HY-14452 5mg Medchemexpress, Fatostatin CAS:125256-00-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY144525MG
Encompass_Preferred
Medchemexpress, HY-14452 5mg Fatostatin CAS:125256-00-0 Fatostatin (125B11), a specific inhibitor of SREBP activation, impairs the activation of SREBP-1 and SREBP-2. Fatostatin binds to SCAP (SREBP cleavage-activating protein), and inhibits the ER-Golgi translocation of SREBPs. Fatostatin decreases the transcription of lipogenic genes in cells. Fatostatin possesses antitumor properties, and lowers hyperglycemia in ob/ob mice[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9051
 Restricted Item