Learn More
Medchemexpress LLC Fatostatin hydrobromide (125B11 hydrobromide) | 298197-04-3 | 99.5% | C18H19BrN2S | 200 MG
SDP
Catalog No. 5000354629
Encompass_Preferred
Fatostatin hydrobromide (125B11 hydrobromide) is a specific inhibitor of SREBP activation that impairs the activation of SREBP-1 and SREBP-2. It binds to SCAP (SREBP cleavage-activating protein) and inhibits the ER-Golgi translocation of SREBPs.
  • Impairs the activation of SREBP-1 and SREBP-2
  • Binds to SCAP (SREBP cleavage-activating protein)
  • Inhibits ER-Golgi translocation of SREBPs
  • Decreases transcription of lipogenic genes in cells
  • Possesses antitumor properties
  • Lowers hyperglycemia in ob/ob mice
  • For research use only
Catalog No. 50-003-54629 Supplier Medchemexpress LLC Supplier No. HY14452A200MG
Fatostatin hydrobromide (125B11 hydrobromide) is a specific inhibitor of SREBP activation that impairs the activation of SREBP-1 and SREBP-2. It binds to SCAP (SREBP cleavage-activating protein) and inhibits the ER-Golgi translocation of SREBPs.
  • Impairs the activation of SREBP-1 and SREBP-2
  • Binds to SCAP (SREBP cleavage-activating protein)
  • Inhibits ER-Golgi translocation of SREBPs
  • Decreases transcription of lipogenic genes in cells
  • Possesses antitumor properties
  • Lowers hyperglycemia in ob/ob mice
  • For research use only