Learn More
TARGETMOL CHEMICALS INC FATOSTATIN HYDROBROMIDE 25MG
SDP
Catalog No. 502576439
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fatostatin hydrobromide (Fatostatin A HBr) ia an inhibitor of sterol regulatory element binding protein (SREBP). It impairs the activation of SREBP-1 and SREBP-2. purity: 99%
Catalog No. 50-257-6439 Supplier TARGETMOL CHEMICALS INC Supplier No. T683225MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fatostatin hydrobromide (Fatostatin A HBr) ia an inhibitor of sterol regulatory element binding protein (SREBP). It impairs the activation of SREBP-1 and SREBP-2. purity: 99%