Learn More
Medchemexpress LLC Fatostatin hydrobromide | 298197-04-3 | MFCD00760867 | 99.52% | C18H19BrN2S | 25 MG
SDP
Catalog No. 5000352998
Encompass_Preferred
Fatostatin hydrobromide (125B11 hydrobromide) is a specific inhibitor of SREBP activation. It impairs the activation of SREBP-1 and SREBP-2 by binding to SCAP (SREBP cleavage-activating protein), which inhibits the ER-Golgi translocation of SREBPs. This action leads to a decrease in the transcription of lipogenic genes in cells.
  • Specific inhibitor of SREBP activation.
  • Impairs activation of SREBP-1 and SREBP-2.
  • Inhibits ER-Golgi translocation of SREBPs.
  • Decreases transcription of lipogenic genes.
  • Exhibits antitumor properties.
  • Lowers hyperglycemia in ob/ob mice.
Catalog No. 50-003-52998 Supplier Medchemexpress LLC Supplier No. 25MGHY14452A
Fatostatin hydrobromide (125B11 hydrobromide) is a specific inhibitor of SREBP activation. It impairs the activation of SREBP-1 and SREBP-2 by binding to SCAP (SREBP cleavage-activating protein), which inhibits the ER-Golgi translocation of SREBPs. This action leads to a decrease in the transcription of lipogenic genes in cells.
  • Specific inhibitor of SREBP activation.
  • Impairs activation of SREBP-1 and SREBP-2.
  • Inhibits ER-Golgi translocation of SREBPs.
  • Decreases transcription of lipogenic genes.
  • Exhibits antitumor properties.
  • Lowers hyperglycemia in ob/ob mice.