Learn More
Medchemexpress LLC HY-14452A 5mg Medchemexpress, Fatostatin (hydrobromide) CAS:298197-04-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14452A5MG
Encompass_Preferred
Medchemexpress, HY-14452A 5mg Fatostatin (hydrobromide) CAS:298197-04-3 Fatostatin hydrobromide (125B11 hydrobromide), a specific inhibitor of SREBP activation, impairs the activation of SREBP-1 and SREBP-2. Fatostatin hydrobromide binds to SCAP (SREBP cleavage-activating protein), and inhibits the ER-Golgi translocation of SREBPs. Fatostatin hydrobromide decreases the transcription of lipogenic genes in cells. Fatostatin hydrobromide possesses antitumor properties, and lowers hyperglycemia in ob/ob mice. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9906