Learn More
Cayman Chemical Fatostatin (hydrobromide), Purity: ≥95%, CAS: 298197-04-3, 50 mg
SDP
Catalog No. 502955371
Encompass_Preferred
An inhibitor of SREBP activation, preventing SCAP-mediated escort of either SREBP-1 or SREBP-2 to the Golgi (IC50 = 5.6 µM); blocks SREBP-mediated gene expression; inhibits high glucose-induced upregulation of TGF-β in primary rat mesangial cells; reduces hepatic fat accumulation in ob/ob mice
Catalog No. 50-295-5371 Supplier Cayman Chemical Supplier No. 1356250 MG
An inhibitor of SREBP activation, preventing SCAP-mediated escort of either SREBP-1 or SREBP-2 to the Golgi (IC50 = 5.6 µM); blocks SREBP-mediated gene expression; inhibits high glucose-induced upregulation of TGF-β in primary rat mesangial cells; reduces hepatic fat accumulation in ob/ob mice