Learn More
Medchemexpress LLC Fatp1-in-2 | 2650944-83-3 | 98.8% | C19H20FN5O | 10MG
SDP
Catalog No. 5000261834
Encompass_Preferred
FATP1-IN-2 is an arylpiperazine-derived small molecule research compound and an orally active inhibitor of fatty acid transport protein 1 (FATP1). It is used in preclinical studies to modulate fatty acid uptake and investigate FATP1-related metabolic processes.
  • Orally active FATP1 inhibitor with reported potency in the low micromolar range.
  • Human FATP1 IC50 = 0.43 μM; mouse FATP1 IC50 = 0.39 μM.
  • High purity (~98.8%) suitable for research applications.
  • Good solubility in DMSO (100 mg/mL); in vivo formulation protocols provided.
  • Stable as a powder at -20°C with defined solvent storage conditions.
  • Available in multiple pack sizes and as a prepared DMSO solution for convenience.
Catalog No. 50-002-61834 Supplier Medchemexpress LLC Supplier No. HY14170010MG
FATP1-IN-2 is an arylpiperazine-derived small molecule research compound and an orally active inhibitor of fatty acid transport protein 1 (FATP1). It is used in preclinical studies to modulate fatty acid uptake and investigate FATP1-related metabolic processes.
  • Orally active FATP1 inhibitor with reported potency in the low micromolar range.
  • Human FATP1 IC50 = 0.43 μM; mouse FATP1 IC50 = 0.39 μM.
  • High purity (~98.8%) suitable for research applications.
  • Good solubility in DMSO (100 mg/mL); in vivo formulation protocols provided.
  • Stable as a powder at -20°C with defined solvent storage conditions.
  • Available in multiple pack sizes and as a prepared DMSO solution for convenience.