Learn More
ABclonal Technology FATP2/SLC27A2 Rabbit mAb
SDP
Catalog No. 5000013137
Encompass_Preferred
The protein encoded by this gene is an isozyme of long-chain fatty-acid-coenzyme A ligase family Although differing in substrate specificity subcellular localization and tissue distribution all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters and thereby play a key role in lipid biosynthesis and fatty acid degradation This isozyme activates long-chain branched-chain and very-long-chain fatty acids containing 22 or more carbons to their CoA derivatives It is expressed primarily in liver and kidney and is present in both endoplasmic reticulum and peroxisomes but not in mitochondria Its decreased peroxisomal enzyme activity is in part responsible for the biochemical pathology in X-linked adrenoleukodystrophy Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-13137 Supplier ABclonal Technology Supplier No. A25501100UL
The protein encoded by this gene is an isozyme of long-chain fatty-acid-coenzyme A ligase family Although differing in substrate specificity subcellular localization and tissue distribution all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters and thereby play a key role in lipid biosynthesis and fatty acid degradation This isozyme activates long-chain branched-chain and very-long-chain fatty acids containing 22 or more carbons to their CoA derivatives It is expressed primarily in liver and kidney and is present in both endoplasmic reticulum and peroxisomes but not in mitochondria Its decreased peroxisomal enzyme activity is in part responsible for the biochemical pathology in X-linked adrenoleukodystrophy Alternatively spliced transcript variants encoding different isoforms have been found for this gene