Learn More
Cusabio Technology LLC fatty acid amide hydrolase
Catalog No. 5000051140
Encompass_Preferred
This kit detects FAAH (Fatty-acid amide hydrolase 1) in rat serum plasma and tissue homogenates using a quantitative sandwich ELISA method with a sensitivity of 0 039 ng/mL and a detection range spanning 0 156 ng/mL to 10 ng/mL The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Absorbance measurements are taken at 450 nm to ensure precise quantification of FAAH levels making it suitable for researchers studying endocannabinoid metabolism or neurological pathways in rat models The optimized protocol provides consistent performance across multiple biological matrices while maintaining high specificity and reproducibility
Catalog No. 50-000-51140 Supplier Cusabio Technology LLC Supplier No. CSBEL007938RA
This kit detects FAAH (Fatty-acid amide hydrolase 1) in rat serum plasma and tissue homogenates using a quantitative sandwich ELISA method with a sensitivity of 0 039 ng/mL and a detection range spanning 0 156 ng/mL to 10 ng/mL The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Absorbance measurements are taken at 450 nm to ensure precise quantification of FAAH levels making it suitable for researchers studying endocannabinoid metabolism or neurological pathways in rat models The optimized protocol provides consistent performance across multiple biological matrices while maintaining high specificity and reproducibility