Learn More
Cayman Chemical Fatty Acid Amide Hydrolase h
SDP
Catalog No. 502888026
Encompass_Preferred
Source: Active recombinant C-terminal His-tagged FAAH expressed in Sf21 cells • Amino acids: 2-579 • MW: 64.3 kDa
Catalog No. 50-288-8026 Supplier Cayman Chemical Supplier No. 10010183100 U
Add to Cart
Add to Cart
Source: Active recombinant C-terminal His-tagged FAAH expressed in Sf21 cells • Amino acids: 2-579 • MW: 64.3 kDa