Learn More
Cayman Chemical Fatty Acid Amide Hydrolase In
SDP
Catalog No. 502886688
Encompass_Preferred
Cayman’s FAAH Inhibitor Screening Assay Kit provides a convenient fluorescence-based method for screening FAAH inhibitors. FAAH hydrolyzes AMC arachidonoyl amide resulting in the release of the fluorescent product, 7-amino-4-methylcoumarin (AMC). The fluorophore can be EAsily analyzed using an excitation wavelength of 340-360 nm and an emission wavelength of 450-465 nm.
Catalog No. 50-288-6688 Supplier Cayman Chemical Supplier No. 1000519696 WELL
Cayman’s FAAH Inhibitor Screening Assay Kit provides a convenient fluorescence-based method for screening FAAH inhibitors. FAAH hydrolyzes AMC arachidonoyl amide resulting in the release of the fluorescent product, 7-amino-4-methylcoumarin (AMC). The fluorophore can be EAsily analyzed using an excitation wavelength of 340-360 nm and an emission wavelength of 450-465 nm.