Learn More
Sigma Aldrich Fine Chemicals Biosciences Fatty Acid Metabolite Libr1EA
Catalog No. 502908342
Encompass
Compounds can either be used as standards and injected individually or a strategy may be employed to pool compounds to allow multiple mixtures to be analyzed per injection.
Catalog No. 50-290-8342 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. FAMLS1EA
Compounds can either be used as standards and injected individually or a strategy may be employed to pool compounds to allow multiple mixtures to be analyzed per injection.