Learn More
Sigma Aldrich Fine Chemicals Biosciences fatty Acid Methyl Esters S
Catalog No. 502904162
Encompass
The fatty Acid Methyl Esters Standard Mixture is a metabolite mixture comprised of 13 fatty acids. fatty acid methyl esters are dissolved in chloroform to the stated concentrations of /- 4%. This product was formulated for use as a retention marker for high-throughput GC/MS metabolomic analysis.
Catalog No. 50-290-4162 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SMB009371ML
The fatty Acid Methyl Esters Standard Mixture is a metabolite mixture comprised of 13 fatty acids. fatty acid methyl esters are dissolved in chloroform to the stated concentrations of /- 4%. This product was formulated for use as a retention marker for high-throughput GC/MS metabolomic analysis.