Learn More
Sigma Aldrich Fine Chemicals Biosciences fatty Acid Methylation Kit
Catalog No. 502904367
Encompass
Methyl esterification of fatty acids is commonly done prior to gas chromatography analysis to prevent peak tailing and to increase sample volatility. However the conventional esterification procedure requires specialized equipment and high technical skill. By using the fatty Acid Methylation kit that employs a new reaction technique followed by the fatty acid methyl ester purification kit (MAK225) fatty acid methyl esterification is greatly simplified.
Catalog No. 50-290-4367 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK2241KT
Methyl esterification of fatty acids is commonly done prior to gas chromatography analysis to prevent peak tailing and to increase sample volatility. However the conventional esterification procedure requires specialized equipment and high technical skill. By using the fatty Acid Methylation kit that employs a new reaction technique followed by the fatty acid methyl ester purification kit (MAK225) fatty acid methyl esterification is greatly simplified.