Learn More
Medchemexpress LLC Fatty Acid Synthase Antibody (YA766) | 273 kDa | 10 UL
SDP
Catalog No. 5000395754
Encompass_Preferred
Fatty Acid Synthase Antibody (YA766) is a mouse-derived, non-conjugated IgG2a monoclonal antibody specifically developed to target and detect Fatty Acid Synthase. This antibody is a valuable tool for research applications in various biological studies.
  • Targets fatty acid synthase
  • Mouse-derived IgG2a monoclonal antibody
  • Suitable for various research applications such as Western Blot (WB), immunocytochemistry/immunofluorescence (ICC/IF), and immunoprecipitation (IP)
Catalog No. 50-003-95754 Supplier Medchemexpress LLC Supplier No. HYP8066810UL
Add to Cart
Add to Cart
Fatty Acid Synthase Antibody (YA766) is a mouse-derived, non-conjugated IgG2a monoclonal antibody specifically developed to target and detect Fatty Acid Synthase. This antibody is a valuable tool for research applications in various biological studies.
  • Targets fatty acid synthase
  • Mouse-derived IgG2a monoclonal antibody
  • Suitable for various research applications such as Western Blot (WB), immunocytochemistry/immunofluorescence (ICC/IF), and immunoprecipitation (IP)