Learn More
ABclonal Technology Fatty Acid Synthase FASN
SDP
Catalog No. 5000007054
Encompass_Preferred
The enzyme encoded by this gene is a multifunctional protein Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA in the presence of NADPH into long-chain saturated fatty acids In some cancer cell lines this protein has been found to be fused with estrogen receptor-alpha (ER-alpha) in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-alpha
Catalog No. 50-000-07054 Supplier ABclonal Technology Supplier No. A22462PM100UL
The enzyme encoded by this gene is a multifunctional protein Its main function is to catalyze the synthesis of palmitate from acetyl-CoA and malonyl-CoA in the presence of NADPH into long-chain saturated fatty acids In some cancer cell lines this protein has been found to be fused with estrogen receptor-alpha (ER-alpha) in which the N-terminus of FAS is fused in-frame with the C-terminus of ER-alpha