Learn More
Abcam Fatty Acid Uptake Assay Kit, 100 Tests
Catalog No. 502205925
Encompass_Preferred
Fatty Acid Uptake Assay Kit (ab287857) enables rapid determination of real-time fatty acid uptake kinetics in live cells expressing fatty acid transporter proteins.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5925 Supplier Abcam Supplier No. AB287857100TEST
Fatty Acid Uptake Assay Kit (ab287857) enables rapid determination of real-time fatty acid uptake kinetics in live cells expressing fatty acid transporter proteins.
The product is subject to the following: Abcam Restricted Use Statement