Learn More
Cayman Chemical Faty ACD DSaturase 1 Monc
SDP
Catalog No. 502940648
Encompass_Preferred
Immunogen: Recombinant human FADS1 protein • Host: Mouse • Species Reactivity: (+) Human; other species not tested • Cross Reactivity: (+) FADS1; (-) FADS2 • Applications: ELISA, IP, and WB • MW = ~45 kDa
Catalog No. 50-294-0648 Supplier Cayman Chemical Supplier No. 27533300 UG
Immunogen: Recombinant human FADS1 protein • Host: Mouse • Species Reactivity: (+) Human; other species not tested • Cross Reactivity: (+) FADS1; (-) FADS2 • Applications: ELISA, IP, and WB • MW = ~45 kDa