Learn More
Cayman Chemical Faty ACD DSaturase 1huma 100ug
SDP
Catalog No. 502934332
Encompass_Preferred
Source: Recombinant N-terminal His-tagged FADS1 N-terminal domain purified from E. coli • Amino acids: 2-121 • MW: 15.89 kDa
Catalog No. 50-293-4332 Supplier Cayman Chemical Supplier No. 25827100 UG
Source: Recombinant N-terminal His-tagged FADS1 N-terminal domain purified from E. coli • Amino acids: 2-121 • MW: 15.89 kDa