Learn More
Cayman Chemical Faty AcId Methyl esTR GCMS 1ea
SDP
Catalog No. 502984481
Encompass_Preferred
A compound that has been used to investigate the relationship between intramolecular rotational dynamics and molecular and crystal structure using NMR spin-lattice relaxation experiments
Catalog No. 50-298-4481 Supplier Cayman Chemical Supplier No. 205031 EA
A compound that has been used to investigate the relationship between intramolecular rotational dynamics and molecular and crystal structure using NMR spin-lattice relaxation experiments