Learn More
Medchemexpress LLC FAUC 213 | 337972-47-1 | 99.9% | C18H19ClN4 | 100 MG
SDP
Catalog No. 5000355539
Encompass_Preferred
FAUC 213 is an orally active and highly selective dopamine D4 receptor complete antagonist. It demonstrates a Ki of 2.2 nM for the hD4.4 receptor and exhibits less activity on D2 and D3 receptors. This compound can cross the blood-brain barrier (BBB) and shows atypical antipsychotic characteristics.
  • Orally active and highly selective dopamine D4 receptor complete antagonist
  • Ki of 2.2 nM for hD4.4 receptor
  • Crosses the blood-brain barrier (BBB)
  • Exhibits atypical antipsychotic characteristics
  • Inhibits p5-HT1 (Ki=1.2 μM), p5-HT2 (Ki=0.52 μM), pα1 (Ki=0.27 μM)
Catalog No. 50-003-55539 Supplier Medchemexpress LLC Supplier No. HY14327100MG
FAUC 213 is an orally active and highly selective dopamine D4 receptor complete antagonist. It demonstrates a Ki of 2.2 nM for the hD4.4 receptor and exhibits less activity on D2 and D3 receptors. This compound can cross the blood-brain barrier (BBB) and shows atypical antipsychotic characteristics.
  • Orally active and highly selective dopamine D4 receptor complete antagonist
  • Ki of 2.2 nM for hD4.4 receptor
  • Crosses the blood-brain barrier (BBB)
  • Exhibits atypical antipsychotic characteristics
  • Inhibits p5-HT1 (Ki=1.2 μM), p5-HT2 (Ki=0.52 μM), pα1 (Ki=0.27 μM)