Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FAUC 213 | 337972-47-1 | 99.9% | C18H19ClN4 | 25 MG
SDP

Catalog No. 5000356494
Encompass_Preferred

FAUC 213 is an orally active and highly selective dopamine D4 receptor complete antagonist with a Ki of 2.2 nM for hD4.4. It shows less activity on D2 and D3 receptors (Kis of 3.4 μM and 5.3 μM for hD2 and hD3, respectively). FAUC 213 can cross the blood-brain barrier (BBB) and exhibits atypical antipsychotic characteristics. It is for research use only and not sold to patients.

  • Targets hD4.4 receptor (Ki = 2.2 nM), hD2 receptor (Ki = 3.4 μM), hD3 receptor (Ki = 5.3 μM)
  • Inhibits p5-HT1 (Ki=1.2 μM), p5-HT2 (Ki=0.52 μM), pα1 (Ki=0.27 μM) in vitro
  • Significantly reduces amphetamine-induced locomotor hyperactivity with pre-treatment of 30 mg/kg in vivo
  • Significantly restores prepulse inhibition reduction caused by apomorphine treatment with 30 mg/kg in vivo

Catalog No. 50-003-56494 Supplier Medchemexpress LLC Supplier No. 25MGHY14327
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FAUC 213 is an orally active and highly selective dopamine D4 receptor complete antagonist with a Ki of 2.2 nM for hD4.4. It shows less activity on D2 and D3 receptors (Kis of 3.4 μM and 5.3 μM for hD2 and hD3, respectively). FAUC 213 can cross the blood-brain barrier (BBB) and exhibits atypical antipsychotic characteristics. It is for research use only and not sold to patients.

  • Targets hD4.4 receptor (Ki = 2.2 nM), hD2 receptor (Ki = 3.4 μM), hD3 receptor (Ki = 5.3 μM)
  • Inhibits p5-HT1 (Ki=1.2 μM), p5-HT2 (Ki=0.52 μM), pα1 (Ki=0.27 μM) in vitro
  • Significantly reduces amphetamine-induced locomotor hyperactivity with pre-treatment of 30 mg/kg in vivo
  • Significantly restores prepulse inhibition reduction caused by apomorphine treatment with 30 mg/kg in vivo

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.