Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FAUC 365 50mg | 474432-66-1 | 462.44 g/mol | C23H25Cl2N3OS | 50 MG
SDP

Catalog No. 5000228169
Encompass_Preferred

FAUC 365 is a dopamine D3 receptor-selective antagonist supplied for preclinical research use. It exhibits high affinity for D3 with markedly lower affinity for D2 and D4 subtypes, and is used in studies of schizophrenia and Parkinson's disease models.

  • High D3 receptor selectivity (reported Ki ≈ 0.5 nM)
  • High purity (~98.8%)
  • Available in solid packs and as a solution in DMSO
  • Soluble in DMSO (~33.33 mg/mL); in vivo formulation guidance provided
  • Recommended storage for powder and solutions to preserve stability
  • Analytical documentation available (SDS, COA, H NMR, RP-HPLC, MS)

Catalog No. 50-002-28169 Supplier Medchemexpress LLC Supplier No. HY11602050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FAUC 365 is a dopamine D3 receptor-selective antagonist supplied for preclinical research use. It exhibits high affinity for D3 with markedly lower affinity for D2 and D4 subtypes, and is used in studies of schizophrenia and Parkinson's disease models.

  • High D3 receptor selectivity (reported Ki ≈ 0.5 nM)
  • High purity (~98.8%)
  • Available in solid packs and as a solution in DMSO
  • Soluble in DMSO (~33.33 mg/mL); in vivo formulation guidance provided
  • Recommended storage for powder and solutions to preserve stability
  • Analytical documentation available (SDS, COA, H NMR, RP-HPLC, MS)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.