Learn More
Medchemexpress LLC FAUC 365 5mg | 474432-66-1 | 462.44 g/mol | C23H25Cl2N3OS | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1160205MG
Encompass_Preferred
FAUC 365 is a research-grade small molecule that acts as a highly selective dopamine D3 receptor antagonist (CAS 474432-66-1). It is supplied as a high-purity solid and is used in biochemical and pharmacological studies related to dopaminergic signaling.
  • Highly selective D3 receptor antagonist with sub-nanomolar affinity (Ki ≈ 0.5 nM)
  • High purity (~98.75%), suitable for in vitro assays
  • Available as a solid and as a 10 mM solution in DMSO for convenience
  • Off-white to light yellow solid, stable under recommended storage
  • Intended for neuroscience research, including schizophrenia and Parkinson's disease models
Catalog No. 50-002-28886