Learn More
Hach Company Faucet Adapter for Portable Softener
Supplier:  Hach Company 1381401
Encompass
Demonstration softener with convenience features. Body of clear polycarbonate plastic. White elliptical base helps prevent upset while in use. Both ends are made of extra thick, white polycarbonate. Quick connect couplets prevent leakage of water. Used with Hach Water Conditioning Demonstration Kit, Model AD-16D.
Catalog No. 50-199-4887