Learn More
Rieke Metals LLC FAUCET DRUM FLO-KING 2" 12/CS
Catalog No. 50371477
Encompass
Two Inch Drum Faucet Flo King, Faucet shoulder & soft durometer gasket,made of EPDM material. Min. order qty. 3 pieces
Catalog No. 50-371-477 Supplier Rieke Metals LLC Supplier No. 04050004
Two Inch Drum Faucet Flo King, Faucet shoulder & soft durometer gasket,made of EPDM material. Min. order qty. 3 pieces