Learn More
New England Biolabs, Inc. FauI – 200 units
SDP
Catalog No. 50994932
Encompass
A restriction endonuclease that recognizes the sequence CCCGCNNNN^NN_.
Catalog No. 50-994-932 Supplier New England Biolabs, Inc. Supplier No. R0651S
A restriction endonuclease that recognizes the sequence CCCGCNNNN^NN_.