Learn More
Selleck Chemical LLC Favipiravir (T-705) 5mg 259793-96-9
SDP
Supplier:  Selleck Chemical LLC S79755MG
Encompass_Preferred
Favipiravir (T-705) is a potent and selective RNA-dependent RNA polymerase inhibitor, used to treat influenza virus infections. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1312443