Learn More
Medchemexpress LLC Fazamorexant | 1808918-69-5 | C25H25FN4O2 | 100 MG
SDP
Catalog No. 5000372969
Encompass_Preferred
Fazamorexant is an orally active dual orexin receptor antagonist with an IC50 of 32 nM against OX1R and 41 nM against OX2R. It prolongs total sleep time and improves sleep efficiency, making it suitable for research on insomnia.
  • Orally active dual orexin receptor antagonist
  • Potently inhibits OX1R (IC50 = 32 nM) and OX2R (IC50 = 41 nM)
  • Prolongs total sleep time
  • Improves sleep efficiency
  • Applicable for research on insomnia
  • Selectively blocks OX1R and OX2R in vivo
  • Promotes rapid onset of sleep by inhibiting wake-promoting neurons
Catalog No. 50-003-72969 Supplier Medchemexpress LLC Supplier No. HY152843100MG
Fazamorexant is an orally active dual orexin receptor antagonist with an IC50 of 32 nM against OX1R and 41 nM against OX2R. It prolongs total sleep time and improves sleep efficiency, making it suitable for research on insomnia.
  • Orally active dual orexin receptor antagonist
  • Potently inhibits OX1R (IC50 = 32 nM) and OX2R (IC50 = 41 nM)
  • Prolongs total sleep time
  • Improves sleep efficiency
  • Applicable for research on insomnia
  • Selectively blocks OX1R and OX2R in vivo
  • Promotes rapid onset of sleep by inhibiting wake-promoting neurons