Learn More
Medchemexpress LLC FB23 | 2243736-35-6 | 98.9% | 377.22 | 50 MG
SDP
Catalog No. 5000351292
Encompass_Preferred
FB23 is a potent and selective FTO demethylase inhibitor with an IC50 of 60 nM. It directly binds to FTO and selectively inhibits FTO's mRNA N6-methyladenosine (m6A) demethylase activity.
  • Potent and selective FTO demethylase inhibitor
  • IC50 of 60 nM against FTO
  • Directly binds to FTO
  • Selectively inhibits FTO's mRNA N6-methyladenosine (m6A) demethylase activity
  • Inhibits proliferation of acute myeloid leukemia (AML) cells (e.g., NB4 and MONOMAC6)
  • Suppresses MYC and E2F targets
  • Activates apoptosis and p53 pathways
Catalog No. 50-003-51292 Supplier Medchemexpress LLC Supplier No. HY13718750MG
FB23 is a potent and selective FTO demethylase inhibitor with an IC50 of 60 nM. It directly binds to FTO and selectively inhibits FTO's mRNA N6-methyladenosine (m6A) demethylase activity.
  • Potent and selective FTO demethylase inhibitor
  • IC50 of 60 nM against FTO
  • Directly binds to FTO
  • Selectively inhibits FTO's mRNA N6-methyladenosine (m6A) demethylase activity
  • Inhibits proliferation of acute myeloid leukemia (AML) cells (e.g., NB4 and MONOMAC6)
  • Suppresses MYC and E2F targets
  • Activates apoptosis and p53 pathways