Learn More
Aviva Systems Biology Corp FBL Antibody - N-terminal region (ARP40366_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40366_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FBL. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human FBL. Alternate names: FIB, FLRN, RNU3IP1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57412