Learn More
Aviva Systems Biology Corp FBLN1 Antibody - N-terminal region (ARP42000_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42000_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBLN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FBLN1. Alternate names: FBLN, FIBL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Yeast
Catalog No. 50-014-58076