Learn More
Medchemexpress LLC FBnG-(Cys-acetamide)-CH2-PEG3-CH2-CH2-CH2-NH2 | 2241669-84-9 | C27H39FN8O6S | 100 MG
SDP
Catalog No. 5000375877
Encompass_Preferred
FBnG-(Cys-acetamide)-CH2-PEG3-CH2-CH2-CH2-NH2 is a tag-linker conjugate of AUTAC4 that incorporates a degradation tag FBnG and a glycol linker. AUTAC4 contains a p-fluorobenzylguanine (FBnG) and a phenylindole moiety, which can induce K63-linked polyubiquitination and degradation of mitochondria in HeLa cells.
  • Tag-linker conjugate for AUTAC4
  • Incorporates a degradation tag FBnG
  • Utilizes a glycol linker
  • Induces K63-linked polyubiquitination
  • Promotes degradation of mitochondria in HeLa cells
  • High purity (98.85%)
  • Appears as a solid
  • White to off-white in color
Catalog No. 50-003-75877 Supplier Medchemexpress LLC Supplier No. HY150408100MG
FBnG-(Cys-acetamide)-CH2-PEG3-CH2-CH2-CH2-NH2 is a tag-linker conjugate of AUTAC4 that incorporates a degradation tag FBnG and a glycol linker. AUTAC4 contains a p-fluorobenzylguanine (FBnG) and a phenylindole moiety, which can induce K63-linked polyubiquitination and degradation of mitochondria in HeLa cells.
  • Tag-linker conjugate for AUTAC4
  • Incorporates a degradation tag FBnG
  • Utilizes a glycol linker
  • Induces K63-linked polyubiquitination
  • Promotes degradation of mitochondria in HeLa cells
  • High purity (98.85%)
  • Appears as a solid
  • White to off-white in color