Learn More
Medchemexpress LLC FBnG-(Cys-acetamide)-CH2-PEG3-CH2-CH2-CH2-NH2 | 2241669-84-9 | C27H39FN8O6S | 25 MG
SDP
Catalog No. 5000374612
Encompass_Preferred
FBnG-(Cys-acetamide)-CH2-PEG3-CH2-CH2-CH2-NH2 is a tag-linker conjugate of AUTAC4 that incorporates a degradation tag FBnG and a glycol linker. AUTAC4 contains an p-fluorobenzylguanine (FBnG) and a phenylindole moiety, which can induce K63-linked polyubiquitination and degradation of mitochondria in HeLa cells.
  • Tag-linker conjugate
  • Incorporates a degradation tag FBnG
  • Glycol linker
  • Induces K63-linked polyubiquitination of mitochondria
  • Induces degradation of mitochondria in HeLa cells
Catalog No. 50-003-74612 Supplier Medchemexpress LLC Supplier No. 25MGHY150408
FBnG-(Cys-acetamide)-CH2-PEG3-CH2-CH2-CH2-NH2 is a tag-linker conjugate of AUTAC4 that incorporates a degradation tag FBnG and a glycol linker. AUTAC4 contains an p-fluorobenzylguanine (FBnG) and a phenylindole moiety, which can induce K63-linked polyubiquitination and degradation of mitochondria in HeLa cells.
  • Tag-linker conjugate
  • Incorporates a degradation tag FBnG
  • Glycol linker
  • Induces K63-linked polyubiquitination of mitochondria
  • Induces degradation of mitochondria in HeLa cells