Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FBnG-(Cys-acetamide)-CH2-PEG3-CH2-CH2-CH2-NH2 | 2241669-84-9 | C27H39FN8O6S | 5 MG
SDP

Catalog No. 5000375223
Encompass_Preferred

FBnG-(Cys-acetamide)-CH2-PEG3-CH2-CH2-CH2-NH2 is a tag-linker conjugate of AUTAC4 that incorporates a degradation tag FBnG and a glycol linker. AUTAC4 contains a p-fluorobenzylguanine (FBnG) and a phenylindole moiety, which can induce K63-linked polyubiquitination and degradation of mitochondria in HeLa cells.

  • Purity: 98.85%
  • Appearance: Solid, white to off-white
  • Molecular weight: 622.71
  • Solubility in DMSO at 50 mg/mL (80.29 mM)
  • For research use only
  • Used in studies related to AUTACs and autophagy

Catalog No. 50-003-75223 Supplier Medchemexpress LLC Supplier No. HY1504085MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FBnG-(Cys-acetamide)-CH2-PEG3-CH2-CH2-CH2-NH2 is a tag-linker conjugate of AUTAC4 that incorporates a degradation tag FBnG and a glycol linker. AUTAC4 contains a p-fluorobenzylguanine (FBnG) and a phenylindole moiety, which can induce K63-linked polyubiquitination and degradation of mitochondria in HeLa cells.

  • Purity: 98.85%
  • Appearance: Solid, white to off-white
  • Molecular weight: 622.71
  • Solubility in DMSO at 50 mg/mL (80.29 mM)
  • For research use only
  • Used in studies related to AUTACs and autophagy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.