Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC FBnG-(Cys-acetamide)-CH2-PEG3-CH2-CH2-CH2-NH2 | 2241669-84-9 | C27H39FN8O6S | 50 MG
SDP

Catalog No. 5000375124
Encompass_Preferred

FBnG-(Cys-acetamide)-CH2-PEG3-CH2-CH2-CH2-NH2 is a tag-linker conjugate of AUTAC4 that incorporates a degradation tag FBnG and a glycol linker. This conjugate can induce K63-linked polyubiquitination and degradation of mitochondria in HeLa cells. For research use only. Store at 4°C, protected from light. In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.

  • Tag-linker conjugate of AUTAC
  • Induces K63-linked polyubiquitination and degradation of mitochondria in HeLa cells
  • Purity: 98.85%

Catalog No. 50-003-75124 Supplier Medchemexpress LLC Supplier No. HY15040850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FBnG-(Cys-acetamide)-CH2-PEG3-CH2-CH2-CH2-NH2 is a tag-linker conjugate of AUTAC4 that incorporates a degradation tag FBnG and a glycol linker. This conjugate can induce K63-linked polyubiquitination and degradation of mitochondria in HeLa cells. For research use only. Store at 4°C, protected from light. In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.

  • Tag-linker conjugate of AUTAC
  • Induces K63-linked polyubiquitination and degradation of mitochondria in HeLa cells
  • Purity: 98.85%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.