Learn More
ABclonal Technology FBP / FOLR1
SDP
Catalog No. 5000005887
Encompass_Preferred
The protein encoded by this gene is a member of the folate receptor family Members of this gene family bind folic acid and its reduced derivatives and transport 5-methyltetrahydrofolate into cells This gene product is a secreted protein that either anchors to membranes via a glycosyl-phosphatidylinositol linkage or exists in a soluble form Mutations in this gene have been associated with neurodegeneration due to cerebral folate transport deficiency Due to the presence of two promoters multiple transcription start sites and alternative splicing multiple transcript variants encoding the same protein have been found for this gene
Catalog No. 50-000-05887 Supplier ABclonal Technology Supplier No. A2241920UL
The protein encoded by this gene is a member of the folate receptor family Members of this gene family bind folic acid and its reduced derivatives and transport 5-methyltetrahydrofolate into cells This gene product is a secreted protein that either anchors to membranes via a glycosyl-phosphatidylinositol linkage or exists in a soluble form Mutations in this gene have been associated with neurodegeneration due to cerebral folate transport deficiency Due to the presence of two promoters multiple transcription start sites and alternative splicing multiple transcript variants encoding the same protein have been found for this gene