Learn More
Aviva Systems Biology Corp FBP2 Antibody - middle region (ARP48089_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48089_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FBP2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human FBP2. Alternate names: MGC142192ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-285-345