Learn More
Medchemexpress LLC FBPase-1 inhibitor-1 | 883973-99-7 | 99.1% | 377.63 | 50 MG
SDP
Catalog No. 5000351573
Encompass_Preferred
FBPase-1 inhibitor-1 (Compound 1) is an allosteric FBPase-1 inhibitor. It blocks gluconeogenesis by inhibiting FBPase-1.
  • IC50 of 3.3 μM or 3.9 μM
  • Can be used for the research of type 2 diabetes (t2dm)
Catalog No. 50-003-51573 Supplier Medchemexpress LLC Supplier No. HY13671750MG
FBPase-1 inhibitor-1 (Compound 1) is an allosteric FBPase-1 inhibitor. It blocks gluconeogenesis by inhibiting FBPase-1.
  • IC50 of 3.3 μM or 3.9 μM
  • Can be used for the research of type 2 diabetes (t2dm)